Gene Transcripts

Gene Symbol
NR1I2
Entrez Gene ID
8856
Gene Name
nuclear receptor subfamily 1 group I member 2
Gene Aliases
BXR, ONR1, PAR, PAR1, PAR2, PARq, PRR, PXR, SAR, SXR
Chromosome Location
Chr. 3: 119780484 - 119818485 on Build GRCh38
UniGene ID
Hs.7303
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003889.3 NP_003880.3 2 1901
NM_022002.2 NP_071285.1 2 227
NM_033013.2 NP_148934.1 2 1901
GenBank mRNA AB307701.1 - 1 62
AF061056.1 - 2 365
AF084644.1 - 2 238
AF084645.1 - 2 341
AJ009936.1 - 3 1901
AJ009937.1 - 3 1901
AK223456.1 - 2 346
AK310789.1 - 2 350
AY091855.1 - 2 644
BC017304.2 - 2 301 5087401
HQ692837.1 - 2 179
HQ709177.1 - 2 179

View Full Product Details