Gene Transcripts

Gene Symbol
Prodh
Entrez Gene ID
19125
Gene Name
proline dehydrogenase
Gene Aliases
Pro-1, Pro1, Ym24d07
Chromosome Location
Chr. 16: 18071726 - 18089283 on Build GRCm38
UniGene ID
Mm.28456
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011172.2 NP_035302.2 10, 11 1289
GenBank mRNA AF120279.1 - 10, 11 1246
AK144068.1 - 10, 11 1259
BC008548.1 - 10, 11 1113
BC125327.1 - 10, 11 1092 40129842
U80020.1 - 10, 11 1238

View Full Product Details