Gene Transcripts

Gene Symbol
Vmn1r49
Entrez Gene ID
24112
Gene Name
vomeronasal 1, receptor 49
Gene Aliases
V1r5, V1rb2, VRi2
Chromosome Location
Chr. 6: 90072086 - 90078374 on Build GRCm38
UniGene ID
Mm.89985
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011911.1 NP_036041.1 1 559
GenBank mRNA AF132114.1 - 1 559

View Full Product Details