Gene Transcripts

Gene Symbol
Dapp1
Entrez Gene ID
26377
Gene Name
dual adaptor for phosphotyrosine and 3-phosphoinositides 1
Gene Aliases
Bam32
Chromosome Location
Chr. 3: 137930998 - 137981576 on Build GRCm38
UniGene ID
Mm.254835
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011932.2 NP_036062.1 9 1633
GenBank mRNA AF186023.1 - 9 1600
AK145745.1 - 9 1599
AK169584.1 - 8 1498

View Full Product Details