Gene Transcripts

Gene Symbol
Anapc7
Entrez Gene ID
56317
Gene Name
anaphase promoting complex subunit 7
Gene Aliases
APC7, AW545589
Chromosome Location
Chr. 5: 122422444 - 122444912 on Build GRCm38
UniGene ID
Mm.37341
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019805.4 NP_062779.3 3 499
GenBank mRNA AF076607.2 - 3 463
AK030775.1 - 3 464
AK049968.1 - 3 438
AK137286.1 - 3 463
AK150764.1 - 3 463
AK150891.1 - 3 463
AK153114.1 - 3 463
AK159523.1 - 3 431
AK164202.1 - 3 468
BC006635.1 - 3 452 3482261

View Full Product Details