Gene Transcripts

Gene Symbol
Nudt3
Entrez Gene ID
56409
Gene Name
nudix (nucleotide diphosphate linked moiety X)-type motif 3
Gene Aliases
1110011B09Rik, AA960325, Dipp, Dipp1
Chromosome Location
Chr. 17: 27579382 - 27623452 on Build GRCm38
UniGene ID
Mm.144699
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291046.1 NP_001277975.1 4 558
NM_019837.2 NP_062811.1 4 550
GenBank mRNA AF264064.1 - 4 549
AK079658.1 - 2 45
BC016534.1 - 5 456 2615904
BC046805.1 - 4 551 6829170
BU919402.1 - 4 382

View Full Product Details