Gene Transcripts

Gene Symbol
Wdr54
Entrez Gene ID
75659
Gene Name
WD repeat domain 54
Gene Aliases
1700030E05Rik
Chromosome Location
Chr. 6: 83152710 - 83156406 on Build GRCm38
UniGene ID
Mm.130665
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023790.2 NP_076279.1 3, 4 491
GenBank mRNA AF053628.1 - 3, 4 479
AK006536.1 - 3, 4 391
AK154345.1 - 3, 4 482
AK202399.1 - 2, 3 106
BC083332.1 - 4, 5 396 6819838

View Full Product Details