Gene Transcripts
- Gene Symbol
- Rab5c
- Entrez Gene ID
- 19345
- Gene Name
- RAB5C, member RAS oncogene family
- Gene Aliases
- AI326010, Rabl
- Chromosome Location
- Chr. 11: 100715009 - 100738210 on Build GRCm38
- UniGene ID
- Mm.29829
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK089199.1 |
- |
6 |
1848 |
|
AK134691.1 |
- |
6 |
1734 |
|
AK167599.1 |
- |
6 |
1783 |
|
View Full Product Details