Gene Transcripts

Gene Symbol
Rnf113a2
Entrez Gene ID
66381
Gene Name
ring finger protein 113A2
Gene Aliases
2310020H19Rik, AI426758, Rnf113a
Chromosome Location
Chr. 12: 84417200 - 84418578 on Build GRCm38
UniGene ID
Mm.24846
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025525.2 NP_079801.2 1 580
GenBank mRNA AK009420.1 - 1 600
AK016795.1 - 1 540
BC058194.1 - 1 486
BC116426.2 - 1 521 40104196

View Full Product Details