Gene Transcripts

Gene Symbol
Metap1d
Entrez Gene ID
66559
Gene Name
methionyl aminopeptidase type 1D (mitochondrial)
Gene Aliases
2310066F24Rik, 3110033D18Rik, AV117938, MAP 1D, Map1d, MetAP 1D, Metapl1
Chromosome Location
Chr. 2: 71453319 - 71525196 on Build GRCm38
UniGene ID
Mm.152796
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025633.3 NP_079909.1 2 166
GenBank mRNA AK005241.1 - 2 163
AK010067.1 - 2 166
BC060739.1 - 2 153

View Full Product Details