Gene Transcripts

Gene Symbol
Nepn
Entrez Gene ID
66650
Gene Name
nephrocan
Gene Aliases
5730521E12Rik, Npn, periolin
Chromosome Location
Chr. 10: 52388870 - 52404611 on Build GRCm38
UniGene ID
Mm.90950
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025684.2 NP_079960.1 2 452
GenBank mRNA AK017656.1 - 2 453
AK017780.1 - 2 453
AK017842.1 - 2 324
AY938536.1 - 2 453
BC132046.1 - 2 442 40130323
BC132048.1 - 2 442 40130325

View Full Product Details