Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000583.3 | NP_000574.2 | 2 | 442 | |
NM_001204306.1 | NP_001191235.1 | 3 | 313 | ||
NM_001204307.1 | NP_001191236.1 | 3 | 250 | ||
XM_006714177.2 | XP_006714240.1 | 2 | 305 | ||
GenBank mRNA | AK223458.1 | - | 3 | 313 | |
AK290827.1 | - | 2 | 152 | ||
AK298433.1 | - | 2 | 156 | ||
AK309595.1 | - | 3 | 250 | ||
AK312971.1 | - | 2 | 156 | ||
AK315853.1 | - | 2 | 305 | ||
BC057228.1 | - | 2 | 160 | 4767205 | |
BX494962.1 | - | 2 | 443 | ||
EU794674.1 | - | 2 | 251 | ||
X03178.1 | - | 2 | 251 |