Gene Transcripts

Gene Symbol
Btbd10
Entrez Gene ID
68815
Gene Name
BTB (POZ) domain containing 10
Gene Aliases
1110056N09Rik, Gmrp1
Chromosome Location
Chr. 7: 113315631 - 113370531 on Build GRCm38
UniGene ID
Mm.238034
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133700.2 NP_598461.1 5 485
GenBank mRNA BC050781.1 - 5 485

View Full Product Details