Gene Transcripts

Gene Symbol
Zdhhc7
Entrez Gene ID
102193
Gene Name
zinc finger, DHHC domain containing 7
Gene Aliases
AL024087, Gramp2
Chromosome Location
Chr. 8: 120081091 - 120101472 on Build GRCm38
UniGene ID
Mm.240076
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133967.3 NP_598728.1 8 1298
GenBank mRNA AK041905.1 - 8 1167
AK089681.1 - 1144
AK090333.1 - 8 1144
AK157968.1 - 8 1151
AK170639.1 - 8 1151
AK170923.1 - 8 1121
AK172462.1 - 8 1132
AY166673.1 - 6 816
BC013467.1 - 7 849 4218025
BC029666.1 - 3 252
BC075666.1 - 8 1298 30605163

View Full Product Details