Gene Transcripts

Gene Symbol
BLVRB
Entrez Gene ID
645
Gene Name
biliverdin reductase B
Gene Aliases
BVRB, FLR, HEL-S-10, SDR43U1
Chromosome Location
Chr. 19: 40447784 - 40465818 on Build GRCh38
UniGene ID
Hs.515785
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000713.2 NP_000704.1 5 659
GenBank mRNA AK312137.1 - 5 586
BC109371.1 - 5 576 5229026
BU528444.1 - 3 254
D26308.1 - 5 607
D32143.1 - 5 638
EU794593.1 - 5 659

View Full Product Details