Gene Transcripts

Gene Symbol
Olfr824
Entrez Gene ID
258669
Gene Name
olfactory receptor 824
Gene Aliases
MOR210-3
Chromosome Location
Chr. 10: 130126108 - 130127055 on Build GRCm38
UniGene ID
Mm.377631
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146674.1 NP_666885.1 1 937
GenBank mRNA BC104094.1 - 1 929
BC104095.2 - 1 929
BC106800.2 - 1 929

View Full Product Details