Gene Transcripts

Gene Symbol
Olfr1408
Entrez Gene ID
258759
Gene Name
olfactory receptor 1408
Gene Aliases
MOR267-4
Chromosome Location
Chr. 1: 173130283 - 173131215 on Build GRCm38
UniGene ID
Mm.389211
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146764.1 NP_666975.1 1 594
GenBank mRNA BC119111.1 - 1 615
BC120665.1 - 1 615 40129588

View Full Product Details