Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001078.3 | NP_001069.1 | 2 | 548 | |
NM_080682.2 | NP_542413.1 | 2 | 548 | ||
GenBank mRNA | AK223266.1 | - | 2 | 447 | |
AK291732.1 | - | 2 | 446 | ||
AU121762.1 | - | 2 | 448 | ||
BC017276.2 | - | 2 | 428 | 4845424 | |
BC068490.1 | - | 2 | 446 | 30339170 | |
BC085003.1 | - | 2 | 428 | 6375943 | |
M30257.1 | - | 2 | 433 | ||
M60335.1 | - | 2 | 426 | ||
X53051.1 | - | 2 | 327 |