Gene Transcripts

Gene Symbol
MOK
Entrez Gene ID
5891
Gene Name
MOK protein kinase
Gene Aliases
RAGE, RAGE-1, RAGE1, STK30
Chromosome Location
Chr. 14: 102224450 - 102305200 on Build GRCh38
UniGene ID
Hs.104119
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001272011.1 NP_001258940.1 3, 4 433
NM_014226.2 NP_055041.1 4, 5 523
XM_006720225.2 XP_006720288.1 4, 5 523
XM_006720226.2 XP_006720289.1 4, 5 523
XM_011537052.1 XP_011535354.1 4, 5 523
XM_017021549.1 XP_016877038.1 4, 5 523
XM_017021550.1 XP_016877039.1 3, 4 422
XM_017021551.1 XP_016877040.1 5, 6 463
XM_017021552.1 XP_016877041.1 4, 5 564
XM_017021555.1 XP_016877044.1 4, 5 523
XM_017021556.1 XP_016877045.1 4, 5 523
XM_017021557.1 XP_016877046.1 3, 4 423
XM_017021558.1 XP_016877047.1 3, 4 389
GenBank mRNA AB022694.1 - 4, 5 517
AK302349.1 - 3, 4 418
AK312778.1 - 4, 5 514
BC026069.1 - 4, 5 513 4838296
BC043179.1 - 1, 2 1115 5287879
BC053536.1 - 4, 5 499 5175851
DB037795.1 - 4, 5 523

View Full Product Details