Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252568.2 | NP_001239497.1 | 3 | 541 | |
NM_001252570.1 | NP_001239499.1 | 2 | 162 | ||
NM_001252571.1 | NP_001239500.1 | 2 | 162 | ||
NM_001281829.1 | NP_001268758.1 | 3 | 188 | ||
NM_172267.4 | NP_758471.1 | 3 | 541 | ||
GenBank mRNA | AI182929.1 | - | 3 | 163 | |
AK005293.1 | - | 3 | 542 | ||
AK046118.1 | - | 2 | 131 | ||
AK138607.1 | - | 3 | 216 | ||
BC039982.1 | - | 3 | 183 | ||
BC132271.1 | - | 3 | 152 | 40130548 | |
BC132273.1 | - | 3 | 152 | 40130550 | |
BC145356.1 | - | 3 | 152 | 9053914 | |
BC171925.1 | - | 3 | 152 | ||
BQ829083.1 | - | 2 | 158 | ||
BY196857.1 | - | 3 | 190 | ||
BY344206.1 | - | 3 | 195 |