Gene Transcripts

Gene Symbol
RPS26
Entrez Gene ID
6231
Gene Name
ribosomal protein S26
Gene Aliases
DBA10, S26
Chromosome Location
Chr. 12: 56041902 - 56044223 on Build GRCh38
UniGene ID
Hs.447562
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029.3 NP_001020.2 2 375
GenBank mRNA AV681946.1 - 2 375
BC002604.1 - 2 339 3143099
BC013215.2 - 1 85
BC015832.1 - 1 136 4807004
BC070220.1 - 1 136 6388128
BC105276.1 - 1 139 4287636
BC105798.1 - 1 140 6726218
X69654.1 - 1 133
X77770.1 - 1 110

Gene Symbol
RPS26P11
Entrez Gene ID
441502
Gene Name
ribosomal protein S26 pseudogene 11
Gene Aliases
RPS26L1, RPS26_22_1784, bA366E13.1
Chromosome Location
Chr. X: 72044409 - 72044974 on Build GRCh38
UniGene ID
Hs.449523
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002309.1 1 246
GenBank mRNA AW972305.1 - 1 356

View Full Product Details