Gene Transcripts

Gene Symbol
Zbbx
Entrez Gene ID
213234
Gene Name
zinc finger, B-box domain containing
Gene Aliases
4931432L23
Chromosome Location
Chr. 3: 75037895 - 75165050 on Build GRCm38
UniGene ID
Mm.80075
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172515.2 NP_766103.2 13 1624
GenBank mRNA AK029899.1 - 13 1617
AK133102.1 - 13 1675
BC050789.1 - 13 1624 6706914

View Full Product Details