Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175124.5 | NP_780333.3 | 10 | 2216 | |
GenBank mRNA | AK077270.1 | - | 10 | 2212 | |
AK159417.1 | - | 10 | 2186 | ||
AK210082.1 | - | 1 | 167 | ||
BC027058.1 | - | 7 | 1774 | ||
BC086694.1 | - | 10 | 2107 | 30697866 |