Gene Transcripts

Gene Symbol
Ralgps1
Entrez Gene ID
241308
Gene Name
Ral GEF with PH domain and SH3 binding motif 1
Gene Aliases
5830418G11Rik, AI853783, AI854138, RALGEF2, RALGPS1A, RalGEF 2, mKIAA0351
Chromosome Location
Chr. 2: 33133419 - 33371511 on Build GRCm38
UniGene ID
Mm.274249
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290570.1 NP_001277499.1 4 492
NM_001290572.1 NP_001277501.1 5 543
NM_175211.5 NP_780420.1 5 543
GenBank mRNA AK076767.1 - 5 528
AK129123.1 - 4 459
AK170248.1 - 5 489
BC072656.1 - 5 478 30536315

View Full Product Details