Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127457.2 | NP_001120929.1 | 9 | 1443 | |
NM_021117.3 | NP_066940.2 | 9 | 1579 | ||
GenBank mRNA | AB014558.1 | - | 9 | 1483 | |
AK291260.1 | - | 9 | 1510 | ||
AK294904.1 | - | 9 | 1437 | ||
AK295627.1 | - | 5 | 979 | ||
AK302865.1 | - | 9 | 1579 | ||
BC035161.1 | - | 11 | 1842 | 5265444 | |
BC041814.1 | - | 9 | 1507 | 5266781 | |
EU219622.1 | - | 9 | 1502 |