Gene Transcripts

Gene Symbol
IL7R
Entrez Gene ID
3575
Gene Name
interleukin 7 receptor
Gene Aliases
CD127, CDW127, IL-7R-alpha, IL7RA, ILRA
Chromosome Location
Chr. 5: 35856875 - 35879603 on Build GRCh38
UniGene ID
Hs.591742
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002185.3 NP_002176.2 6 831
GenBank mRNA AK315251.1 - 6 730
BC020717.1 - 6 817
BC067537.1 - 6 808 6971840
BC067538.1 - 6 808 6971842
BC067539.1 - 6 808 6971844
BC067540.1 - 6 808 6971846
BC069999.1 - 6 808 6971843
M29696.1 - 6 750

View Full Product Details