Gene Transcripts

Gene Symbol
MSI1
Entrez Gene ID
4440
Gene Name
musashi RNA binding protein 1
Gene Aliases
Chromosome Location
Chr. 12: 120339655 - 120369180 on Build GRCh38
UniGene ID
Hs.158311
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002442.3 NP_002433.1 10 772
XM_006719403.3 XP_006719466.1 10 766
XM_006719404.3 XP_006719467.1 5 551
XM_011538361.2 XP_011536663.1 10 766
XM_011538362.2 XP_011536664.1 10 766
XM_011538363.2 XP_011536665.1 10 766
XM_011538364.2 XP_011536666.1 6 1316
XM_011538365.2 XP_011536667.1 7 435
XM_011538366.2 XP_011536668.1 7 1443
XM_011538368.2 XP_011536670.1 5 391
XM_011538370.2 XP_011536672.1 5 317
XM_011538371.2 XP_011536673.1 5 343
GenBank mRNA AB012851.1 - 10 747
AB072590.1 - 7 494
AB072591.1 - 7 494
AB073212.1 - 7 487
AK023262.1 - 3 192
BX393884.2 - 10 773

View Full Product Details