Gene Transcripts

Gene Symbol
PRKCZ
Entrez Gene ID
5590
Gene Name
protein kinase C zeta
Gene Aliases
PKC-ZETA, PKC2
Chromosome Location
Chr. 1: 2050470 - 2185395 on Build GRCh38
UniGene ID
Hs.496255
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033581.1 NP_001028753.1 5, 6 643
NM_001033582.1 NP_001028754.1 5, 6 540
NM_001242874.1 NP_001229803.1 5, 6 650
NM_002744.4 NP_002735.3 8, 9 855
XM_011541773.1 XP_011540075.1 8, 9 851
XM_011541774.1 XP_011540076.1 5, 6 716
XM_011541775.2 XP_011540077.1 8, 9 851
XM_011541776.1 XP_011540078.1 5, 6 656
XM_011541778.1 XP_011540080.1 5, 6 569
XM_017001789.1 XP_016857278.1 8, 9 851
XM_017001790.1 XP_016857279.1 8, 9 851
XM_017001791.1 XP_016857280.1 5, 6 500
XM_017001792.1 XP_016857281.1 5, 6 656
XM_017001793.1 XP_016857282.1 5, 6 914
XM_017001794.1 XP_016857283.1 5, 6 569
XM_017001795.1 XP_016857284.1 5, 6 573
XM_017001796.1 XP_016857285.1 5, 6 656
XM_017001797.1 XP_016857286.1 3, 4 297
XM_017001798.1 XP_016857287.1 5, 6 569
XM_017001799.1 XP_016857288.1 5, 6 977
XM_017001800.1 XP_016857289.1 3, 4 297
XM_017001801.1 XP_016857290.1 3, 4 297
XM_017001802.1 XP_016857291.1 5, 6 853
XM_017001803.1 XP_016857292.1 8, 9 851
XM_017001804.1 XP_016857293.1 5, 6 763
XM_017001805.1 XP_016857294.1 5, 6 733
GenBank mRNA AK097627.1 - 3, 4 276
AK290995.1 - 8, 9 694
AK294649.1 - 5, 6 645
AK294782.1 - 5, 6 563
AK302160.1 - 3, 4 267
AK310372.1 - 3, 4 297
BC008058.2 - 8, 9 802 2987996
BC014270.2 - 8, 9 855 3835020
BT007082.1 - 8, 9 694
L14283.1 - 8, 9 694
Z15108.1 - 8, 9 676

View Full Product Details