Gene Transcripts

Gene Symbol
TAF5
Entrez Gene ID
6877
Gene Name
TATA-box binding protein associated factor 5
Gene Aliases
TAF(II)100, TAF2D, TAFII-100, TAFII100
Chromosome Location
Chr. 10: 103367953 - 103389065 on Build GRCh38
UniGene ID
Hs.96103
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006951.4 NP_008882.2 3 976
NM_139052.2 NP_620640.1 3 976
GenBank mRNA AK223615.1 - 3 948
AK291229.1 - 3 976
BC052268.1 - 3 954 6575596
BC136340.1 - 3 957 9020327
BC136348.1 - 3 957 9020335
U75309.1 - 3 942
U80191.1 - 3 953
X95525.1 - 3 962

View Full Product Details