Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001171162.1 | NP_001164633.1 | 24 | 4607 | |
NM_005096.3 | NP_005087.1 | 24 | 4082 | ||
NM_201599.2 | NP_963893.1 | 24 | 4073 | ||
XM_005262309.3 | XP_005262366.1 | 24 | 4516 | ||
XM_005262310.2 | XP_005262367.1 | 24 | 3993 | ||
XM_011531062.2 | XP_011529364.1 | 24 | 3982 | ||
GenBank mRNA | AB002383.1 | - | 24 | 3981 | |
BC069057.1 | - | 24 | 3986 | 5499547 | |
X95808.1 | - | 25 | 4583 |