Gene Transcripts
- Gene Symbol
- SH3PXD2A
- Entrez Gene ID
- 9644
- Gene Name
- SH3 and PX domains 2A
- Gene Aliases
- FISH, SH3MD1, TKS5
- Chromosome Location
- Chr. 10: 103594027 - 103855464 on Build GRCh38
- UniGene ID
- Hs.678727
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014631.2 |
NP_055446.2
|
10, 11 |
974 |
|
XM_005270294.4 |
XP_005270351.1
|
11, 12 |
1107 |
|
XM_005270295.4 |
XP_005270352.1
|
10, 11 |
1061 |
|
XM_005270297.3 |
XP_005270354.1
|
5, 6 |
2338 |
|
XM_005270298.4 |
XP_005270355.1
|
5, 6 |
1141 |
|
XM_006718079.3 |
XP_006718142.1
|
12, 13 |
1204 |
|
XM_006718080.3 |
XP_006718143.1
|
6, 7 |
1235 |
|
XM_011540385.2 |
XP_011538687.1
|
11, 12 |
1158 |
|
XM_011540386.2 |
XP_011538688.1
|
6, 7 |
2434 |
|
XM_011540387.2 |
XP_011538689.1
|
2, 3 |
4080 |
|
XM_017016969.1 |
XP_016872458.1
|
11, 12 |
1125 |
|
XM_017016970.1 |
XP_016872459.1
|
10, 11 |
941 |
|
GenBank mRNA |
AB007878.1 |
- |
8, 9 |
724 |
|
AK056469.1 |
- |
9, 10 |
697 |
|
AK294454.1 |
- |
5, 6 |
514 |
|
AK294736.1 |
- |
5, 6 |
552 |
|
AK295314.1 |
- |
5, 6 |
591 |
|
AK295660.1 |
- |
5, 6 |
618 |
|
AK302015.1 |
- |
8, 9 |
730 |
|
AK304828.1 |
- |
5, 6 |
530 |
|
AK315983.1 |
- |
5, 6 |
557 |
|
View Full Product Details