Gene Transcripts

Gene Symbol
ST6GALNAC2
Entrez Gene ID
10610
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 2
Gene Aliases
SAITL1, SIAT7, SIAT7B, SIATL1, ST6GalNAII, STHM
Chromosome Location
Chr. 17: 76565379 - 76586066 on Build GRCh38
UniGene ID
Hs.592105
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006456.2 NP_006447.2 5, 6 923
XM_005256954.3 XP_005257011.1 5, 6 926
GenBank mRNA AJ251053.1 - 5, 6 707
AK225943.1 - 5, 6 721
BC040455.1 - 5, 6 705 4252792
BT019972.1 - 5, 6 668
U14550.1 - 5, 6 707

View Full Product Details