Gene Transcripts

Gene Symbol
SLITRK3
Entrez Gene ID
22865
Gene Name
SLIT and NTRK like family member 3
Gene Aliases
Chromosome Location
Chr. 3: 165186720 - 165197109 on Build GRCh38
UniGene ID
Hs.101745
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318810.1 NP_001305739.1 2 1010
NM_001318811.1 NP_001305740.1 2 663
NM_014926.3 NP_055741.2 2 848
GenBank mRNA AB020655.1 - 2 677
AK290024.1 - 2 1010
BC114621.1 - 2 662 40036558

View Full Product Details