Gene Transcripts

Gene Symbol
ACIN1
Entrez Gene ID
22985
Gene Name
apoptotic chromatin condensation inducer 1
Gene Aliases
ACINUS, ACN, fSAP152
Chromosome Location
Chr. 14: 23058564 - 23095614 on Build GRCh38
UniGene ID
Hs.124490
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164814.1 NP_001158286.1 15, 16 3447
NM_001164815.1 NP_001158287.1 14, 15 3366
NM_001164816.1 NP_001158288.1 8, 9 1005
NM_001164817.1 NP_001158289.1 9, 10 1067
NM_014977.3 NP_055792.1 15, 16 3486
XM_005267415.3 XP_005267472.1 15, 16 3287
XM_005267416.3 XP_005267473.1 15, 16 3254
XM_005267418.1 XP_005267475.1 8, 9 1302
GenBank mRNA AB014570.2 - 15, 16 2995
AF124726.1 - 15, 16 3485
AF124727.1 - 9, 10 1021
AF124728.1 - 8, 9 996
AK299034.1 - 14, 15 2877
AL050382.1 - 8, 9 2091
AL833228.1 - 10, 11 1336
BC140805.1 - 15, 16 3216 9021673
BX247975.1 - 8, 9 1275

View Full Product Details