Gene Transcripts

Gene Symbol
KIRREL
Entrez Gene ID
55243
Gene Name
kin of IRRE like (Drosophila)
Gene Aliases
NEPH1
Chromosome Location
Chr. 1: 157993273 - 158100262 on Build GRCh38
UniGene ID
Hs.272234
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286349.1 NP_001273278.1 13 2050
NM_018240.6 NP_060710.3 15 2350
XM_005245305.4 XP_005245362.1 15 2055
GenBank mRNA AK001707.1 - 8 1042
AK022708.1 - 11 1400
AK297788.1 - 13 1678
AY302131.1 - 13 2045
BC025268.1 - 10 1246
BC051356.1 - 8 1013
BC067097.1 - 8 1013
BC082969.1 - 8 1013
BC109192.1 - 15 2013 40001501
BC109193.1 - 15 2013 40001502

View Full Product Details