Gene Transcripts

Gene Symbol
UEVLD
Entrez Gene ID
55293
Gene Name
UEV and lactate/malate dehyrogenase domains
Gene Aliases
ATTP, UEV3
Chromosome Location
Chr. 11: 18529609 - 18588837 on Build GRCh38
UniGene ID
Hs.407991
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040697.2 NP_001035787.1 5 555
NM_001261382.1 NP_001248311.1 4 489
NM_001261383.1 NP_001248312.1 4 489
NM_001261384.1 NP_001248313.1 3 306
NM_001261385.1 NP_001248314.1 5 512
NM_001297771.1 NP_001284700.1 5 555
NM_018314.4 NP_060784.3 5 555
XM_017017985.1 XP_016873474.1 4 482
XM_017017986.1 XP_016873475.1 4 362
GenBank mRNA AF503350.1 - 5 515
AK001930.1 - 5 512
AK295501.1 - 3 234
AK296835.1 - 4 449
AK314521.1 - 5 555
AK316191.1 - 4 362
BC011011.1 - 5 492
BC064566.1 - 4 429 5576632
CB991602.1 - 5 570

View Full Product Details