Gene Transcripts

Gene Symbol
PGBD5
Entrez Gene ID
79605
Gene Name
piggyBac transposable element derived 5
Gene Aliases
Chromosome Location
Chr. 1: 230321646 - 230425928 on Build GRCh38
UniGene ID
Hs.520463
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258311.1 NP_001245240.1 4 938
XM_017002330.1 XP_016857819.1 4 2007
GenBank mRNA AL833243.1 - 3 583
BC036865.1 - 3 365
KF670820.1 - 4 739

View Full Product Details