Gene Transcripts

Gene Symbol
TEX35
Entrez Gene ID
84066
Gene Name
testis expressed 35
Gene Aliases
C1orf49, TSC24
Chromosome Location
Chr. 1: 178513077 - 178523500 on Build GRCh38
UniGene ID
Hs.534501
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170722.1 NP_001164193.1 2 187
NM_001170723.1 NP_001164194.1 2 163
NM_001170724.1 NP_001164195.1 2 163
NM_032126.4 NP_115502.2 2 163
GenBank mRNA AK093269.1 - 2 110
AL136694.1 - 2 167
BC034972.2 - 2 120 4823687
BI461686.1 - 2 128
BI825610.1 - 2 97

View Full Product Details