Gene Transcripts

Gene Symbol
SSMEM1
Entrez Gene ID
136263
Gene Name
serine rich single-pass membrane protein 1
Gene Aliases
C7orf45
Chromosome Location
Chr. 7: 130205399 - 130216869 on Build GRCh38
UniGene ID
Hs.351816
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145268.3 NP_660311.1 3 304
XM_005250141.3 XP_005250198.1 3 211
XM_011515794.2 XP_011514096.1 3 168
XM_011515795.2 XP_011514097.1 3 161
GenBank mRNA AK097635.1 - 3 290
BC017587.2 - 3 308 4838989

View Full Product Details