Gene Transcripts

Gene Symbol
AMOT
Entrez Gene ID
154796
Gene Name
angiomotin
Gene Aliases
Chromosome Location
Chr. X: 112774877 - 112840946 on Build GRCh38
UniGene ID
Hs.528051
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113490.1 NP_001106962.1 10 3116
NM_133265.2 NP_573572.1 11 2685
XM_005262087.1 XP_005262144.1 13 3861
XM_005262090.1 XP_005262147.1 10 2358
XM_011530875.2 XP_011529177.1 14 3973
XM_017029289.1 XP_016884778.1 12 2889
GenBank mRNA AB028994.3 - 9 1889
AF286598.1 - 11 2685
AY987378.1 - 10 3116
BC130294.1 - 9 2109 40146325

View Full Product Details