Gene Transcripts
- Gene Symbol
- TAPT1
- Entrez Gene ID
- 202018
- Gene Name
- transmembrane anterior posterior transformation 1
- Gene Aliases
- CMVFR, OCLSBG
- Chromosome Location
- Chr. 4: 16160505 - 16226538 on Build GRCh38
- UniGene ID
- Hs.479223
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153365.2 |
NP_699196.2
|
9, 10 |
1179 |
|
XM_005248139.2 |
XP_005248196.1
|
8, 9 |
993 |
|
XM_005248140.3 |
XP_005248197.1
|
8, 9 |
943 |
|
XM_011513812.2 |
XP_011512114.1
|
8, 9 |
1042 |
|
XM_011513815.2 |
XP_011512117.1
|
8, 9 |
2278 |
|
XM_011513816.2 |
XP_011512118.1
|
8, 9 |
987 |
|
XM_011513817.2 |
XP_011512119.1
|
6, 7 |
556 |
|
XM_017007876.1 |
XP_016863365.1
|
10, 11 |
1069 |
|
GenBank mRNA |
AK074494.1 |
- |
9, 10 |
1112 |
|
AK296104.1 |
- |
8, 9 |
981 |
|
AK299163.1 |
- |
5, 6 |
475 |
|
BC066899.1 |
- |
9, 10 |
1117 |
4825556 |
View Full Product Details