Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001031679.2 | NP_001026849.1 | 6 | 636 | |
NM_001193460.1 | NP_001180389.1 | 7 | 800 | ||
NM_001193461.1 | NP_001180390.1 | 6 | 498 | ||
NM_198080.3 | NP_932346.1 | 5 | 509 | ||
GenBank mRNA | AK293084.1 | - | 6 | 606 | |
AK299065.1 | - | 5 | 448 | ||
AK316365.1 | - | 5 | 480 | ||
AL833622.1 | - | 6 | 554 | ||
AY358229.1 | - | 5 | 444 | ||
BC040053.1 | - | 7 | 701 | 6025598 | |
BX500552.1 | - | 6 | 500 | ||
BX648776.1 | - | 6 | 555 | ||
DC357690.1 | - | 5 | 509 |