Gene Transcripts

Gene Symbol
TRIM42
Entrez Gene ID
287015
Gene Name
tripartite motif containing 42
Gene Aliases
PPP1R40, T4A1
Chromosome Location
Chr. 3: 140678024 - 140701150 on Build GRCh38
UniGene ID
Hs.343487
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152616.4 NP_689829.3 3 1949
XM_011512740.2 XP_011511042.1 3 1944
GenBank mRNA AF521868.1 - 3 1761
AK097416.1 - 3 1936
BC130472.1 - 3 1859 40146503

View Full Product Details