Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175856.4 | NP_787052.3 | 3 | 1839 | |
XM_011543363.2 | XP_011541665.1 | 3 | 1692 | ||
XM_011543364.2 | XP_011541666.1 | 2 | 1048 | ||
XM_017009434.1 | XP_016864923.1 | 2 | 10738 | ||
GenBank mRNA | AB086062.1 | - | 3 | 1839 | |
AB175496.1 | - | 3 | 1839 | ||
AJ504664.2 | - | 3 | 1376 | ||
AJ578034.2 | - | 3 | 2197 | ||
AY726604.1 | - | 3 | 1141 | ||
BC137325.1 | - | 3 | 1908 | 9021322 |