Gene Transcripts

Gene Symbol
SEPN1
Entrez Gene ID
57190
Gene Name
selenoprotein N, 1
Gene Aliases
CFTD, MDRS1, RSMD1, RSS, SELN
Chromosome Location
Chr. 1: 25800176 - 25818222 on Build GRCh38
UniGene ID
Hs.323396
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020451.2 NP_065184.2 7, 8 1066
NM_206926.1 NP_996809.1 6, 7 964
GenBank mRNA AF166125.1 - 5, 6 655
AJ306399.1 - 7, 8 1066
AK308457.1 - 5, 6 684
BC021028.2 - 2, 3 269
BQ217758.1 - 4, 5 521

View Full Product Details