Gene Transcripts

Gene Symbol
SNHG12
Entrez Gene ID
85028
Gene Name
small nucleolar RNA host gene 12
Gene Aliases
ASLNC04080, C1orf79, LINC00100, NCRNA00100
Chromosome Location
Chr. 1: 28578538 - 28581854 on Build GRCh38
UniGene ID
Hs.632377
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024127.1 1 12
GenBank mRNA AK092096.1 - 1 33
BC045792.1 - 1 30
BC068048.1 - 1 12
KJ782215.1 - 2 1141

View Full Product Details