Gene Transcripts

Gene Symbol
RPPH1
Entrez Gene ID
85495
Gene Name
ribonuclease P RNA component H1
Gene Aliases
H1RNA, RPPH1-1
Chromosome Location
Chr. 14: 20343071 - 20343411 on Build GRCh38
UniGene ID
Hs.613102
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002312.1 1 83
GenBank mRNA CA413366.1 - 1 278
DW429001.1 - 1 99
X15624.1 - 1 82

Gene Symbol
PARP2
Entrez Gene ID
10038
Gene Name
poly(ADP-ribose) polymerase 2
Gene Aliases
ADPRT2, ADPRTL2, ADPRTL3, ARTD2, PARP-2, pADPRT-2
Chromosome Location
Chr. 14: 20343041 - 20357904 on Build GRCh38
UniGene ID
Hs.409412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017020912.1 XP_016876401.1 1 285

View Full Product Details