Gene Transcripts

Gene Symbol
MRPL21
Entrez Gene ID
219927
Gene Name
mitochondrial ribosomal protein L21
Gene Aliases
L21mt, MRP-L21
Chromosome Location
Chr. 11: 68891276 - 68903835 on Build GRCh38
UniGene ID
Hs.503047
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181514.1 NP_852615.1 5 448
NM_181515.1 NP_852616.1 5 461
XM_005273823.4 XP_005273880.1 5 445
GenBank mRNA AK096756.1 - 5 461
AK301989.1 - 5 445
BC055088.1 - 5 447 6457142
BM916618.1 - 5 413

View Full Product Details