Gene Transcripts

Gene Symbol
LST1
Entrez Gene ID
7940
Gene Name
leukocyte specific transcript 1
Gene Aliases
B144, D6S49E, LST-1
Chromosome Location
Chr. 6: 31586179 - 31588909 on Build GRCh38
UniGene ID
Hs.436066
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166538.1 NP_001160010.1 3 277
NM_007161.3 NP_009092.3 4 370
NM_205837.2 NP_995309.2 4 456
NM_205838.2 NP_995310.2 4 393
NM_205839.2 NP_995311.2 5 486
NM_205840.2 NP_995312.2 3 326
NR_029461.1 3 370
NR_029462.1 3 325
XM_006715206.3 XP_006715269.1 5 499
XM_006715209.3 XP_006715272.1 4 402
XM_006715210.3 XP_006715273.1 4 457
XM_011514914.2 XP_011513216.1 5 480
GenBank mRNA AF000424.1 - 4 456
AF000425.1 - 5 464
AF000426.1 - 3 325
AI091696.1 - 1 280
AJ705986.1 - 3 307
AV711621.1 - 3 289
AV715235.1 - 4 448
AY852265.1 - 3 181
AY852266.1 - 4 270
BC103855.1 - 3 261 40003000
BC103856.1 - 3 261 40003004
BI838080.1 - 4 306
BX647293.1 - 2 1552
CD742958.1 - 1 292
DQ099382.1 - 3 228
Y18486.1 - 3 278
Y18487.1 - 3 326
Y18488.1 - 3 211
Y18489.1 - 2 254
Y18490.1 - 2 188

View Full Product Details