Gene Transcripts
- Gene Symbol
- SPATA31D4
- Entrez Gene ID
- 389761
- Gene Name
- SPATA31 subfamily D member 4
- Gene Aliases
- FAM75D4
- Chromosome Location
- Chr. 9: 81928428 - 81934998 on Build GRCh38
- UniGene ID
- Hs.722630
- Species
- Homo sapiens
- Gene Symbol
- SPATA31D3
- Entrez Gene ID
- 389762
- Gene Name
- SPATA31 subfamily D member 3
- Gene Aliases
- FAM75D3
- Chromosome Location
- Chr. 9: 81943500 - 81950094 on Build GRCh38
- UniGene ID
- Hs.522246
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207416.2 |
NP_997299.2
|
4 |
1271 |
|
View Full Product Details